Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Broth w/Polysorbate 80

Catalog No. R112755 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 90 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R112755 10 x 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R112755 Supplier Thermo Scientific™ Supplier No. R112755
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel Tryptic Soy Broth w/Polysorbate 80 is used for sterility testing.

Thermo Scientific™ Remel Tryptic Soy Broth w/Polysorbate 80 is used for sterility testing.

TRUSTED_SUSTAINABILITY

Specifications

Description Tryptic Soy Broth w/Polysorbate 80
Format WMB Lug - 90mL
Product Type Microbiology Media
Quantity 10 x 90 mL

Not intended for IVD use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.