Learn More
Thermo Scientific™ Urea Agar
GSA_VA
Catalog No. R065212 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100/Pk.
20/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R065212 20/Pk.
R065210 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R065212 Supplier Thermo Scientific™ Supplier No. R065212
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Detect urease activity and differentiate microorganisms on the basis of urease production using Urea Agar.
Differentiate microorganisms on the basis of urease activity with Thermo Scientific™ Remel™ Urea Agar. Christensen developed this medium to help in differentiation of rapid urea-positive Proteus spp. from other urea-positive enteric gram-negative bacilli1,2. This medium allows for urease activity to be demonstrated in organisms which have more complex nitrogen requirements3. The medium can also be used to determine urease activity of non-enteric gram-negative bacilli, such as Brucella and Bordetella and to differentiate Cryptococcus and other yeasts4,5.
Urea Agar is used to detect rapid urease activity of the Proteae and non-rapid urease activity of some Enterobacteriaceae. Urea Agar is a lightly buffered medium that contains urea and phenol red, a pH indicator. When organisms utiliz
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM.
Description Urea Agar
Format 15mm x 103mm Tube - Slant
Product Type Agar Slants
Quantity 20/Pk.