Description
Differentiate genera within the Enterobacteriaceae on the basis of urease production with Thermo Scientific™ Remel™ Urea Broth (Stuart's). Urea Broth (Stuart's) was developed by Stuart et al. to differentiate Proteus from other genera within the Enterobacteriaceae1,2. This medium is designed such that it detects urease activity in organisms which produce ammonia in a highly buffered medium with limited nutrients. Such organisms include Proteus spp., Morganella morganii, Providencia rettgeri, and urease-positive Providencia stuartii3.
Urea Broth (Stuart’s) is a liquid medium used to detect microorganisms on the basis of urease production. It is a medium with high buffering capacity and limited nutrients. Other genera within the Enterobacteriaceae which hydrolyze urea, will not
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Urea Broth |
| Format | 15mm x 103mm Tube |
| Product Type | Microbiology Media |
| Quantity | 20 x 2 mL |