Learn More
Description
Isolate and identify Gardnerella vaginalis with this differential, ready-to-use solid medium.
Isolate and identify Gardnerella vaginalis with Thermo Scientific™ Remel™ V Agar prepared plates, a medium containing a special blood agar base with 5% human blood, designed to enhance the growth of fastidious organisms.
Use the solid media V Agar Monoplate to isolate and identify Gardnerella vaginalis.
- Promotes: V Agar contains the essential elements for the growth of Gardnerella vaginalis.
- Easy to read: The transparent agar allows differentiation of G. vaginalis colonies from those of commensal microbial flora (e.g., streptococci, yeasts, lactobacilli, etc.).
- Selective: Colistin, nalidixic acid and nystatin inhibit most gram-negative bacilli, yeast and diptheroids.
- Ready-to-use: Convenience of prepared media.
Casein and meat peptones supply nitrogenous compounds, carbon, sulphur and trace elements necessary for the growth of G. vaginalis. Cornstarch is an energy source. Human blood is incorporated into the agar to allow for presumptive identification of G. vaginalis, which produces diffuse beta-hemolysis on V agar and Agar Selective. Colistin, nalidixic acid and nystatin are selective agents, which inhibit most gram-negative bacilli, yeast and diptheroids.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | V Agar |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
