Learn More
Thermo Scientific™ V Agar Selective (w/colistin, nalidixic acid, nystatin)
Shop All Thermo Scientific ProductsDescription
Isolate and presumptively identify Gardnerella vaginalis from clinical specimens using these prepared plates.
Isolate and presumptively identify Gardnerella vaginalis from clinical specimens using Thermo Scientific™ Remel™ V Agar Selective (w/colistin, nalidixic acid, nystatin) prepared plates, a selective medium containing a special blood agar base designed to enhance the growth of fastidious organisms while maintaining osmotic balance.
Gardnerella vaginalis is found in the genitourinary tract of females and is an indicator organism for bacterial vaginosis (BV). BV is a risk factor for premature labor, chorioamnionitis, and postpartum endometritis. Women with postpartum fever and neonates with sepsis may be infected with Gardnerella vaginalis. In males, Gardnerella vaginalis has been associated with urinary tract infections, prostatitis, and urethritis. Gardnerella vaginalis is a gram-variable rod or coccobacillus. Beta hemolysis is observed on human or rabbit blood-containing media, but not sheep blood agars.
- Selective: selective agents colistin, nalidixic acid, nystatin inhibit most Gram-negative bacilli, yeasts and diphtheroid.
- Ready to use convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
Specifications
Specifications
| Description | V Agar; Selective w/colistin, nalidixic acid, nystatin |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.