Learn More
Thermo Scientific™ VACC Agar (w/vancomycin amphotericin B, ceftazidime, and clindamycin)
Catalog No. R01954 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01954 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01954 Supplier Thermo Scientific™ Supplier No. R01954
Add to Cart
Add to Cart
This item is not returnable. View return policy
Carry out primary, selective isolation of extended-spectrum-beta-lactamase producing Enterobacteriaceae with these prepared plates.
Carry out primary, selective isolation of extended-spectrum-beta-lactamase producing Enterobacteriaceae in qualitative procedures with Thermo Scientific™ Remel™ VACC Agar (w/vancomycin amphotericin B, ceftazidime, and clindamycin).
Extended Spectrum β Lactamases are enzymes capable of conferring bacterial resistance to penicillins, cephalosporins, and aztreonam by hydrolysis of these antibiotics 1,2. Production of ESBLs is a significant resistance-mechanism that impedes the antimicrobial treatment of infections caused by Enterobacteriaceae and is a serious threat to the currently available antibiotic armory3.
The medium can facilitate earlier identification of colonized patients and can help reduce the cost of surveillance screening.
  • Selective: selective agents vancomycin, amphotericin B, ceftazidime, and clindamycin inhibit many commensal organisms found in the large intestine while at the same time allowing the growth of certain resistant organisms.
  • Facilitates earlier identification of colonized patients and can help reduce the cost of surveillance screening.
  • Ready to use convenience of prepared media.
In a study conducted from 2000 to 2005, there were 17,872 inpatients screened for rectal colonization with ESBL-producing Enterobacteriaceae. The colonization rate doubled during this study4.During the same study cited above, 8.5% of colonized patients developed a subsequent bloodstream infection4. Surveillance screening is useful for infection control and antimicrobial stewardship in controlling the proliferation of ESBL-producing bacteria. The use of VACC Agar can facilitate earlier identification of colonized patients and can help reduce the cost of surveillance screening.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
TRUSTED_SUSTAINABILITY
Specifications
Description VACC Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.