Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Vitamin K, Hemin Solution

Catalog No. R450951 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R450951 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R450951 Supplier Thermo Scientific™ Supplier No. R450951
Add to Cart
Add to Cart
This item is not returnable. View return policy

Remel Vitamin K, Hemin Solution is used in anaerobic media.

Remel Vitamin K, Hemin Solution is used in anaerobic media.

TRUSTED_SUSTAINABILITY

Specifications

Concentration Vitamin K: 0.5mg/10mL. Hemin: 5mg/10mL.
Description Vitamin K, Hemin Solution
Form Liquid
Product Type Solution
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.