Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Voges-Proskauer B Test
GSA_VA

Catalog No. R21281 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21281 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21281 Supplier Thermo Scientific™ Supplier No. R21281
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Voges-Proskauer B Test.

Determine the ability of gram-negative bacilli to produce acetylmethylcarbinol (acetoin) from glucose fermentation using Thermo Scientific™ Remel™ 40% Potassium Hydroxide Reagent for Voges-Proskauer B Test. In 1898, Voges and Proskauer reported production of acetylmethylcarbinol from glucose fermentation by certain bacteria when grown in an appropriate medium such as MR-VP Broth1,2. Later Barret made the VP test more sensitive by using 40% Potassium Hydroxide (VP-B) to the medium for the detection of acetylmethylcarbinol production3.

The Voges-Proskauer test is based on the detection of acetylmethylcarbinol from glucose metabolism4.Ease of Differentiation: VP positive organisms produce a pink color development while VP negative organisms produce no color.Rapid Detection: Within 15 minutes.Glucose is metabolized to pyruvic acid from which the neutral products acetoin and 2,3-butanediol are formed. In the presence of oxygen and alkali, acetoin and 2,3-butanediol are oxidized to diacetyl, the reactant for the pink color produced in Voges-Proskauer test. VP-B is an oxidizing agent which hastens oxidation of acetoin to diacetyl.Used in conjunction with VP-A, Part No.R21200. For use with MRVP Broth.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at Room Temperature Until Used
Description Voges-Proskauer B
Detectable Analytes Acetylmethylcarbinol
Packaging Type 25 mL/Bottle
Quantity 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.