Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Xanthine Agar

Catalog No. R09974 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R09974 20 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R09974 Supplier Thermo Scientific™ Supplier No. R09974

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate aerobic actinomycetes on the basis of xanthine hydrolysis using Xanthine Agar.

Differentiate aerobic actinomycetes on the basis of xanthine hydrolysis using Thermo Scientific™ Remel™ Xanthine Agar. Mishra et al. developed a taxonomic scheme for aerobic actinomycetes on the basis of biochemical characteristics1. A simplified scheme for identification of most commonly isolated actinomycetes was developed by McGinnis et al. and includes tyrosine, xanthine, hypoxanthine, and casein tests2.

The aerobic acinomycetes are a heterogeneous group of genera and include the species of Nocardia and Streptomyces3. As a group, the organisms are gram-positive, catalase positive, and may be presumptively identified by staining procedures, microscopic morphology, and substrate degradation.

  • Ready-to-use – Convenience of a prepared media
  • Easy-to-interpret – Clearing of xanthine due to enzymatic hydrolysis around and/or under growth indicates a positive test. Growth without clearing of medium indicates a negative test.

This medium contains beef extract and peptone that provide carbohydrates, vitamins, nitrogen compounds and salts essential for the growth of aerobic actinomycetes. Peptone also provides nutrients in the form of amino acids and peptides. This medium detects the presence of the hydrolytic enzyme that degrades xanthine resulting in clearing of the media surrounding and beneath the areas of growth.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Xanthine Agar
Format G - PT - 25mL
Product Type Agar
Quantity 20 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.