Learn More
Thermo Scientific™ Flu A and B Control Swab
GSA_VA
Catalog No. R246003 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
40 Swabs
Catalog No. Quantity
R246003 40 Swabs
1 options
This item is not returnable. View return policy
Catalog No. R246003 Supplier Thermo Scientific™ Supplier No. R246003
Add to Cart
Add to Cart
This item is not returnable. View return policy
Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Flu A and B test kits with Thermo Scientific Xpect Remel Flu A and B Control Swab.
Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Flu A and B test kits with Thermo Scientific Xpect Remel Flu A and B Control Swab.
These kits contain positive and negative controls to monitor test performance.
Human influenza A and B viruses cause seasonal epidemics of disease (known as the flu season) almost every winter. Flu A is known to cause flu pandemics. The Influenza A and B test is performed for the qualitative detection and differentiation of Influenza A virus, including H5N1 and H1N1, and Influenza B virus in nasal/throat/nasopharyngeal swab or nasopharyngeal aspirate.
This kit contains Flu A control dry swab containing inactivated influenza A antigen and Flu B control dry swab containing inactivated influenza B antigen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description Xpect Flu A and B Control Swab
Detectable Analytes Influenza
Type Control
Includes 20 Flu A+/Flu B- control swabs and 20 Flu A-/Flu B+ control swabs
Quantity 40 Swabs