Learn More
Thermo Scientific™ Remel™ Xpect™ Giardia/Cryptosporidium Control Kit
GSA_VA
Catalog No. 23313103 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3 Vials/Kit
Catalog No. Quantity
23-313-103 3 Vials/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-313-103 Supplier Thermo Scientific™ Supplier No. R2450530
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use in qualitative procedures as external control material for the Xpect Cryptosporidium, Xpect Giardia, or Xpect Giardia/Cryptosporidium test kits.
Remel Xpect Giardia/Cryptosporidium Control Kit is used in qualitative procedures as external control material for the Xpect Cryptosporidium, Xpect Giardia, or Xpect Giardia/Cryptosporidium test kits.
Includes:
1 Giardia positive vial, 1 Cryptosporidium positive vial and 1 negative vial
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description Xpect™ Giardia/Cryptosporidium Control Kit
CE Marker Yes
Detectable Analytes Giardia and Cryptosporidium Antigen
Quantity 3 Vials/Kit