Learn More
Thermo Scientific™ Rotavirus Control Kit
Catalog No. R246551 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 Vials/Kit
Catalog No. Quantity
R246551 2 Vials/Kit
1 options
This item is not returnable. View return policy
Catalog No. R246551 Supplier Thermo Scientific™ Supplier No. R246551
Add to Cart
Add to Cart
This item is not returnable. View return policy
Monitor gross systematic failures using Xpect Rotavirus Control Kit as external quality control material for Xpect Rotavirus Kit.
Monitor gross systematic failures using Thermo Scientific™ Remel™ Xpect™ Rotavirus Control Kit as external quality control material for Xpect Rotavirus Kit. This kit contains a negative and a positive control.
Monitor gross systematic failures using Xpect Rotavirus Control Kit as external quality control material for Xpect Rotavirus Kit.
  • Ready to Use
  • Used as a confirmatory test before the patient sample is used.
  • Should be tested according to Xpect Rotavirus Instructions For Use (IFU).
This kit is used for qualitative procedures. The test should produce negative and positive results with the corresponding control before testing the patient sample.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description Control Kit
Includes 1mL/Vial positive control; 1mL/Vial negative control
Quantity 2 Vials/Kit