Learn More
Thermo Scientific™ Remel™ Xpect™ RSV Control Kit
Catalog No. R246012 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 Vials
Catalog No. Quantity
R246012 2 Vials
1 options
This item is not returnable. View return policy
Catalog No. R246012 Supplier Thermo Scientific™ Supplier No. R246012
Add to Cart
Add to Cart
This item is not returnable. View return policy
Remel Xpect RSV Control Kit is an Xpect RSV liquid control set.
Remel Xpect RSV Control Kit is an Xpect RSV liquid control set.
  • Method: Chromatographic immunoassay
  • Samples collected from nasopharyngeal specimens in neonatal and paediatric patients
Ordering Alerts:
Six positive and negative tests can be run from 1 set of controls.
Includes:
1 vial positive control and 1 vial negative control. 1mL/Vial.
TRUSTED_SUSTAINABILITY
Specifications
Description 6 Tests
Type Control
Includes 1 Vial positive control and 1 Vial negative control. 1mL/Vial.
Quantity 2 Vials