Learn More
Medchemexpress LLC HY-100365 100mg Medchemexpress, Remetinostat CAS:946150-57-8 Purity:>98%
SDP
Catalog No. 501931788
Encompass_Preferred
Medchemexpress, HY-100365 100mg Remetinostat CAS:946150-57-8 Remetinostat (SHP-141) is a hydroxamic acid-based inhibitor of histone deacetylase enzymes (HDAC) which is under development for the treatment of cutaneous T-cell lymphoma [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1788 Supplier Medchemexpress LLC Supplier No. HY100365100MG
Medchemexpress, HY-100365 100mg Remetinostat CAS:946150-57-8 Remetinostat (SHP-141) is a hydroxamic acid-based inhibitor of histone deacetylase enzymes (HDAC) which is under development for the treatment of cutaneous T-cell lymphoma [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.