Learn More
AdipoGen Remisporine B (2.5 mg)
SDP
Supplier:  AdipoGen AGCN20179MM25
Encompass_Preferred
Chemical. CAS: 571194-06-4. Formula: C30H24O12. MW: 576.5. Isolated from fungus Penicillium sp. Antibiotic. Mycotoxin. Exhibits very weak in vitro antimicrobial activity, as well as weak cytotoxic and immunosuppressive activity. Remisporine B is a unique cyclopentachromone (CPC) dimer resulting from a spontaneous Diels-Alder reaction of Remisporine A.
Catalog No. 50-205-3570