Learn More
Medchemexpress LLC Remodelin hydrobromide | 1622921-15-6 | 99.6% | 363.28 | 1 ML
SDP
Catalog No. 5000390545
Encompass_Preferred
Remodelin hydrobromide is an orally active and selective inhibitor of acetyltransferase NAT10, which inhibits NAT10 activity and slows DNA replication.
  • Suppresses the growth of prostate cancer cells.
  • Inhibits the growth of prostate cancer and hepatocellular carcinoma in xenograft models.
  • Enhances the healthspan in a Hutchinson-Gilford progeria syndrome (HGPS) mouse model.
Catalog No. 50-003-90545 Supplier Medchemexpress LLC Supplier No. HY16706A10MM/1ML
Remodelin hydrobromide is an orally active and selective inhibitor of acetyltransferase NAT10, which inhibits NAT10 activity and slows DNA replication.
  • Suppresses the growth of prostate cancer cells.
  • Inhibits the growth of prostate cancer and hepatocellular carcinoma in xenograft models.
  • Enhances the healthspan in a Hutchinson-Gilford progeria syndrome (HGPS) mouse model.