Learn More
TARGETMOL CHEMICALS INC REMODELIN HYDROBROMIDE 25MG
SDP
Catalog No. 502579530
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Remodelin hydrobromide (Remodelin HBR) is a novel potent and selective inhibitor of the acetyl-transferase protein NAT10. purity: 99%
Catalog No. 50-257-9530 Supplier TARGETMOL CHEMICALS INC Supplier No. T613325MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Remodelin hydrobromide (Remodelin HBR) is a novel potent and selective inhibitor of the acetyl-transferase protein NAT10. purity: 99%