Learn More
Cayman Chemical RemodelInhydrobromIde 10mg
SDP
Catalog No. 502971389
Encompass_Preferred
A 2-thiazolylhydrazone derivative that, at 10 µM, improves nuclear architecture, chromatin organization, and survival of both cells lacking lamin A and cells from patients with progeria; inhibits NAT10
Catalog No. 50-297-1389 Supplier Cayman Chemical Supplier No. 1606610 MG
A 2-thiazolylhydrazone derivative that, at 10 µM, improves nuclear architecture, chromatin organization, and survival of both cells lacking lamin A and cells from patients with progeria; inhibits NAT10