Learn More
Cayman Chemical Remodelin (hydrobromide), Purity: ≥98%, CAS: 1622921-15-6, 5 mg
SDP
Catalog No. 502973123
Encompass_Preferred
A 2-thiazolylhydrazone derivative that, at 10 µM, improves nuclear architecture, chromatin organization, and survival of both cells lacking lamin A and cells from patients with progeria; inhibits NAT10
Catalog No. 50-297-3123 Supplier Cayman Chemical Supplier No. 160665 MG
A 2-thiazolylhydrazone derivative that, at 10 µM, improves nuclear architecture, chromatin organization, and survival of both cells lacking lamin A and cells from patients with progeria; inhibits NAT10