Learn More
Cayman Chemical Remogliflozin A, Purity: ≥98%, CAS: 329045-45-6, 10 mg
SDP
Catalog No. 502952667
Encompass_Preferred
A potent inhibitor of SGLT2 (Kis = 12.4 and 26 nM for human and rat SGLT2, respectively); selective for SGLT2 over SGLT1 (Kis = 4,520 and 997 nM for human and rat SGLT1, respectively)
Catalog No. 50-295-2667 Supplier Cayman Chemical Supplier No. 1434010 MG
A potent inhibitor of SGLT2 (Kis = 12.4 and 26 nM for human and rat SGLT2, respectively); selective for SGLT2 over SGLT1 (Kis = 4,520 and 997 nM for human and rat SGLT1, respectively)