Learn More
Medchemexpress LLC Remogliflozin etabonate | 442201-24-3 | 99.6% | 522.59 | 100 MG
SDP
Catalog No. 5000360220
Encompass_Preferred
Remogliflozin etabonate is an orally active, selective, and low-affinity sodium glucose cotransporter (SGLT2) inhibitor. It acts as a prodrug, converting to its active form, Remogliflozin, within the body. This compound has demonstrated antidiabetic efficacy in various rodent models, offering potential benefits in glucose and lipid metabolism regulation.
  • Orally active and selective SGLT2 inhibitor
  • Functions as a prodrug, metabolized to active Remogliflozin
  • Exhibits antidiabetic efficacy in rodent models
  • Significantly reduces fasting plasma glucose (FPG) and glycated hemoglobin (GHb) levels
  • Ameliorates hyperglycemia and increases urinary glucose excretion
  • Decreases plasma insulin in normal rats
  • Suppresses the development of hypertriglyceridemia
Catalog No. 50-003-60220 Supplier Medchemexpress LLC Supplier No. HY14945100MG
Remogliflozin etabonate is an orally active, selective, and low-affinity sodium glucose cotransporter (SGLT2) inhibitor. It acts as a prodrug, converting to its active form, Remogliflozin, within the body. This compound has demonstrated antidiabetic efficacy in various rodent models, offering potential benefits in glucose and lipid metabolism regulation.
  • Orally active and selective SGLT2 inhibitor
  • Functions as a prodrug, metabolized to active Remogliflozin
  • Exhibits antidiabetic efficacy in rodent models
  • Significantly reduces fasting plasma glucose (FPG) and glycated hemoglobin (GHb) levels
  • Ameliorates hyperglycemia and increases urinary glucose excretion
  • Decreases plasma insulin in normal rats
  • Suppresses the development of hypertriglyceridemia