Learn More
Medchemexpress LLC Remogliflozin etabonate | 442201-24-3 | 99.6% | 522.59 | 50 MG
SDP
Catalog No. 5000360312
Encompass_Preferred
Remogliflozin etabonate is an orally active, selective, and low-affinity sodium glucose cotransporter (SGLT2) inhibitor. It acts as a proagent, based on benzylpyrazole glucoside, and is metabolized in the body to its active form, Remogliflozin. This compound has demonstrated antidiabetic efficacy in rodent models.
  • Orally active and selective SGLT2 inhibitor
  • Metabolized to active form, Remogliflozin
  • Demonstrates antidiabetic efficacy in rodent models
  • Ki values for hSGLT2, rSGLT2, hSGLT1, and rSGLT1
Catalog No. 50-003-60312 Supplier Medchemexpress LLC Supplier No. HY1494550MG
Remogliflozin etabonate is an orally active, selective, and low-affinity sodium glucose cotransporter (SGLT2) inhibitor. It acts as a proagent, based on benzylpyrazole glucoside, and is metabolized in the body to its active form, Remogliflozin. This compound has demonstrated antidiabetic efficacy in rodent models.
  • Orally active and selective SGLT2 inhibitor
  • Metabolized to active form, Remogliflozin
  • Demonstrates antidiabetic efficacy in rodent models
  • Ki values for hSGLT2, rSGLT2, hSGLT1, and rSGLT1