Learn More
Caron Products And Services REMOTE ALARM CONTACTS FOR 7000
Catalog No. NC1976379
Encompass_Preferred
Remote Alarm Contacts for 7000 Series Chambers. Provides NO and NC dry contacts for alarms.
Catalog No. NC1976379 Supplier Caron Products And Services Supplier No. ALRM302
Remote Alarm Contacts for 7000 Series Chambers. Provides NO and NC dry contacts for alarms.