Learn More
Research Products International Corp Remote Temperature/Humidity Monitor with Simultaneous Display with Certificate
SDP
Catalog No. NC1586164
Encompass
Ideal for monitoring temperature, humidity or combination of both inside a desiccator, refrigerator, freezer or any controlled environment chamber. Each model features a high accuracy mounted to a six foot long cable. Allows convenient placement of both sensor and monitor. Monitor includes a built in stand and can also be wall mounted. Can easily be moved to different locations.
Supplied with AAA battery.
Catalog No. NC1586164 Supplier Research Products International Corp Supplier No. 800027C
Add to Cart
Add to Cart
Ideal for monitoring temperature, humidity or combination of both inside a desiccator, refrigerator, freezer or any controlled environment chamber. Each model features a high accuracy mounted to a six foot long cable. Allows convenient placement of both sensor and monitor. Monitor includes a built in stand and can also be wall mounted. Can easily be moved to different locations.
Supplied with AAA battery.