Learn More
Research Products International Corp Remote Temperature/Humidity Monitor with Simultaneous Display
SDP
Catalog No. 50999191
Encompass
Ideal for monitoring temperature, humidity or combination of both inside a desiccator, refrigerator, freezer or any controlled environment chamber. Each model features a high accuracy mounted to a six foot long cable. Allows convenient placement of both sensor and monitor. Monitor includes a built in stand and can also be wall mounted. Can easily be moved to different locations.
Supplied with AAA battery.
Catalog No. 50-999-191 Supplier Research Products International Corp Supplier No. 800027
Ideal for monitoring temperature, humidity or combination of both inside a desiccator, refrigerator, freezer or any controlled environment chamber. Each model features a high accuracy mounted to a six foot long cable. Allows convenient placement of both sensor and monitor. Monitor includes a built in stand and can also be wall mounted. Can easily be moved to different locations.
Supplied with AAA battery.