Learn More
Grainger Remote Test Station 2 Watt
Catalog No. 502766104
Encompass_Preferred
The Edwards Signaling Remote Test Station is a 2-watt remote test station that allows for testing of fire alarm systems. It features a red Lexan test switch and plate, and includes a DPDT switch to activate alarm circuits. The test station has screw terminal connections and operates on 24VDC. It is made of durable ABS plastic with a red finish and measures 5.2" H x 4" W x 2.75" D. The remote test station is UL Listed and can be surface or flush mounted for convenient access during fire alarm system testing.
Catalog No. 50-276-6104 Supplier Grainger Supplier No. 16X441
The Edwards Signaling Remote Test Station is a 2-watt remote test station that allows for testing of fire alarm systems. It features a red Lexan test switch and plate, and includes a DPDT switch to activate alarm circuits. The test station has screw terminal connections and operates on 24VDC. It is made of durable ABS plastic with a red finish and measures 5.2" H x 4" W x 2.75" D. The remote test station is UL Listed and can be surface or flush mounted for convenient access during fire alarm system testing.