Learn More
Grainger Remote Test Station 4 Watt
Catalog No. 502766658
Encompass_Preferred
The Edwards Signaling Remote Test Station is a 4-watt remote test station that allows for testing of fire alarm notification appliances. It features a red enclosure with a horn strobe and provides a remote testing point for notification appliances, eliminating the need to activate the panel alarm. This UL Listed device has a maximum sound level of 88 dB at 10 feet and provides visual and audible notification. The remote test station operates at 12 or 24 volts DC and is compatible with System Sensor, Wheelock, and Gentex devices. It enables weekly testing per NFPA 72 requirements.
Catalog No. 50-276-6658 Supplier Grainger Supplier No. 16X442
The Edwards Signaling Remote Test Station is a 4-watt remote test station that allows for testing of fire alarm notification appliances. It features a red enclosure with a horn strobe and provides a remote testing point for notification appliances, eliminating the need to activate the panel alarm. This UL Listed device has a maximum sound level of 88 dB at 10 feet and provides visual and audible notification. The remote test station operates at 12 or 24 volts DC and is compatible with System Sensor, Wheelock, and Gentex devices. It enables weekly testing per NFPA 72 requirements.