Learn More
Grainger Edwards Signaling Remote Zone Indicator, 10 Zone, White
Catalog No. 502766205
Encompass_Preferred
The Remote Zone Indicator is a 10-zone annunciator designed for use with E-FSC1004 fire alarm control panels. This unit provides status indication for up to ten distinct zones, featuring a white finish and a versatile design suitable for flush or surface mounting.
  • Provides ten zones of annunciation
  • Compatible with E-FSC1004 control panels
  • Supports 24 VDC operation
  • Designed for indoor use within a temperature range of 32°F to 120°F (0°C to 49°C)
  • Features a 98 mA alarm current and 17,000 μA standby current
  • Constructed to meet UL, ULC, and CSFM standards
  • Dimensions: 8 1/2 in. (length) x 5 5/8 in. (height) x 1 1/2 in. (depth)
Catalog No. 50-276-6205 Supplier Grainger Supplier No. 16X401
The Remote Zone Indicator is a 10-zone annunciator designed for use with E-FSC1004 fire alarm control panels. This unit provides status indication for up to ten distinct zones, featuring a white finish and a versatile design suitable for flush or surface mounting.
  • Provides ten zones of annunciation
  • Compatible with E-FSC1004 control panels
  • Supports 24 VDC operation
  • Designed for indoor use within a temperature range of 32°F to 120°F (0°C to 49°C)
  • Features a 98 mA alarm current and 17,000 μA standby current
  • Constructed to meet UL, ULC, and CSFM standards
  • Dimensions: 8 1/2 in. (length) x 5 5/8 in. (height) x 1 1/2 in. (depth)