Learn More
Grainger Remote Zone Indicator 98mA White
Catalog No. 502766202
Encompass_Preferred
The Kidde Remote Zone Indicator is a 10-zone remote indicator panel for use with Kidde intelligent addressable fire alarm control panels. It provides LED indications for up to 10 addressable zones, with each zone having an associated LED that illuminates red during an alarm condition and yellow during a trouble condition. The panel features a compact design with dimensions of 10" x 6" x 1.25" and can be surface or semi-flush mounted. It connects to the fire alarm control panel via an included ribbon cable and gets its power from the panel, with no additional power supply required. This provides an easy way to remotely annunciate addressable zone status for up to 10 zones.
Catalog No. 50-276-6202 Supplier Grainger Supplier No. 16X402
The Kidde Remote Zone Indicator is a 10-zone remote indicator panel for use with Kidde intelligent addressable fire alarm control panels. It provides LED indications for up to 10 addressable zones, with each zone having an associated LED that illuminates red during an alarm condition and yellow during a trouble condition. The panel features a compact design with dimensions of 10" x 6" x 1.25" and can be surface or semi-flush mounted. It connects to the fire alarm control panel via an included ribbon cable and gets its power from the panel, with no additional power supply required. This provides an easy way to remotely annunciate addressable zone status for up to 10 zones.