Learn More
GA International Removable Blackout Labels 0.9" x 0.75" / 22.9mm x 19.1mm Rectangle White From -20°C to +100°C (-4°F to +212°F)
Catalog No. 5000571820
Encompass_Preferred
Cover existing information with these easily removable blackout labels made to fit microscope slides. Ideal for re-identifying slides marked with unclear, erroneous, or incomplete information. These waterproof labels are made with a durable thermoplastic film and leave behind no residue when removed. Available in roll format and are intended for use with all major brands of thermal-transfer printers.
Catalog No. 50-005-71820 Supplier GA International Supplier No. AZA152NPC36WH
Cover existing information with these easily removable blackout labels made to fit microscope slides. Ideal for re-identifying slides marked with unclear, erroneous, or incomplete information. These waterproof labels are made with a durable thermoplastic film and leave behind no residue when removed. Available in roll format and are intended for use with all major brands of thermal-transfer printers.