Learn More
GA International Removable Cryogenic Labels for Laser Printers 1.28" x 0.5" / 32.5mm x 12.7mm Rectangle Green Apple From -196°C to +120°C (-321°F to +248°F)
Catalog No. 5000571066
Encompass_Preferred
Cryogenic thermoplastic laser labels with a glove-friendly removable adhesive. Ideal for long-term storage of samples in liquid nitrogen (on flat surfaces such as polycarbonate cryo boxes) and harsh deep-freeze environments (as low as -80°C; on vials, tubes, freezer boxes, microplates, and others) used in biomedical research and clinical laboratories. Cryo labels accept desktop laser printing and permanent markers. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.
Catalog No. 50-005-71066 Supplier GA International Supplier No. RCL23T1GA
Cryogenic thermoplastic laser labels with a glove-friendly removable adhesive. Ideal for long-term storage of samples in liquid nitrogen (on flat surfaces such as polycarbonate cryo boxes) and harsh deep-freeze environments (as low as -80°C; on vials, tubes, freezer boxes, microplates, and others) used in biomedical research and clinical laboratories. Cryo labels accept desktop laser printing and permanent markers. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.