Learn More
GA International Removable Cryogenic Labels for Laser Printers 2.64" x 0.277" / 67.1mm x 7mm Rectangle Red From -196°C to +120°C (-321°F to +248°F)
Catalog No. 5000572619
Encompass_Preferred
Cryogenic thermoplastic laser labels with a glove-friendly removable adhesive. Ideal for long-term storage of samples in liquid nitrogen (on flat surfaces such as polycarbonate cryo boxes) and harsh deep-freeze environments (as low as -80°C; on vials, tubes, freezer boxes, microplates, and others) used in biomedical research and clinical laboratories. Cryo labels accept desktop laser printing and permanent markers. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.
Catalog No. 50-005-72619 Supplier GA International Supplier No. RCL11T1RE
Cryogenic thermoplastic laser labels with a glove-friendly removable adhesive. Ideal for long-term storage of samples in liquid nitrogen (on flat surfaces such as polycarbonate cryo boxes) and harsh deep-freeze environments (as low as -80°C; on vials, tubes, freezer boxes, microplates, and others) used in biomedical research and clinical laboratories. Cryo labels accept desktop laser printing and permanent markers. Water-resistant; wear and tear proof; do not jam printers; resistant to UV and gamma irradiation.