Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CoolCell™ FTS30 Vial Module
BIOPHARMA

Catalog No. 07210078 Shop All Corning Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-210-078 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-210-078 Supplier Corning™ Supplier No. 432078
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with CoolCell™ FTS30. Corning CoolCell™ FTS30 Vial Module is a cryogenic vials holder that allows one-step insertion and removal of all 30 vials at once.

Corning CoolCell FTS30 Vial Module is a holder for 30 1.0 mL or 2.0 mL cryogenic vials that allows one-step insertion and removal of all 30 vials at once. It fits into a standard 5.0 x 5.0 x 2.0 inch cryostorage box and is compatible with dry ice and liquid nitrogen.
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Application) Holding cryogenic vials
No. per Case 10
No. per Pack 10
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.