Learn More
GA International Removable Deep-Freeze Labels for Thermal-Transfer Barcode Printers 1" x 0.6875" / 25.4mm x 17.5mm Rectangle Yellow From -80°C to +100°C (-112°F to +212°F)
Catalog No. 5000571012
Encompass_Preferred
Deep-freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (-20°C, -40°C, -80°C) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.Waterproof and tearproof, these removable deep-freeze labels can also withstand temperatures ranging up to +100°C, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.
Catalog No. 50-005-71012 Supplier GA International Supplier No. RMTT75NPC31YE
Deep-freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (-20°C, -40°C, -80°C) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.Waterproof and tearproof, these removable deep-freeze labels can also withstand temperatures ranging up to +100°C, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.