Learn More
GA International Removable Deep-Freeze Thermal-Transfer Labels for Microplates 2" x 0.275" / 50.8mm x 7mm Rectangle Blue From -80°C to +110°C (-112°F to +230°F)
Catalog No. 5000572133
Encompass_Preferred
Glove-friendly, removable deep-freeze labels designed to identify 96-well and 384-well, skirted and semi-skirted microtiter plates (microplates) intended for safe use in PCR thermocyclers and sequencing instruments. These versatile labels can be stored in low-temperature conditions such as refrigerators (4°C/39°F), laboratory freezers (-80°C/-112°F), and dry ice transportation; they can also withstand temperatures up to 110°C (230°F). Supplied in roll format on 1” or 3” cores, they are compatible with all major brands of thermal-transfer printers.
Catalog No. 50-005-72133 Supplier GA International Supplier No. AMA223C12BL
Add to Cart
Add to Cart
Glove-friendly, removable deep-freeze labels designed to identify 96-well and 384-well, skirted and semi-skirted microtiter plates (microplates) intended for safe use in PCR thermocyclers and sequencing instruments. These versatile labels can be stored in low-temperature conditions such as refrigerators (4°C/39°F), laboratory freezers (-80°C/-112°F), and dry ice transportation; they can also withstand temperatures up to 110°C (230°F). Supplied in roll format on 1” or 3” cores, they are compatible with all major brands of thermal-transfer printers.