Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Removable Hose Connection, For Condensers
GREENER_CHOICESDP

Catalog No. 31501980 Shop All Kemtech Products
Change view
Click to view available options
Thread Size:
Size #14
Size #18
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size
31-501-980 Size #18
31-501-979 Size #14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-501-980 Supplier Synthware™ Supplier No. R201801
Add to Cart
Add to Cart

Replacement hose connections - Straight

Specifications

Color Red
For Use With (Application) Organic Synthesis
Includes No hole
Material Plastic
Product Type Hose Connection
Quantity 1 Ea.
Thread Size Size #18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.