Learn More
Trajan Scientific REMOVABLE NEEDLE SYRINGE
SDP
Catalog No. 502530356
Encompass_Preferred
Trajan Autosampler Syringes with Removable Needle for Bruker (Varian) CP-8400/8410, CP-9010/9050, Connection Type: Removable, Diameter Outer Needle: 0.018 in., 0.47 mm, Needle Gauge: 26 G, Model: 10R-BT-GT-0.47C, Length Needle: 50 mm, 1.9 in., Volume: 5 uL
Catalog No. 50-253-0356 Supplier Trajan Scientific Supplier No. 0029851
Trajan Autosampler Syringes with Removable Needle for Bruker (Varian) CP-8400/8410, CP-9010/9050, Connection Type: Removable, Diameter Outer Needle: 0.018 in., 0.47 mm, Needle Gauge: 26 G, Model: 10R-BT-GT-0.47C, Length Needle: 50 mm, 1.9 in., Volume: 5 uL