Learn More
World Precision Instrument Removable Zero Point
SDP
Catalog No. 502539188
Encompass
  • Probe Centering Device mounts on the “U” frame of all WPI Rat Stereotaxic Instruments, and allows replacing and centering the probe (for use only with ear bar at 0 coordinates), without removing the animal.
  • The hole is 30 mm above ear bar zero.
Catalog No. 50-253-9188 Supplier World Precision Instrument Supplier No. 505224
  • Probe Centering Device mounts on the “U” frame of all WPI Rat Stereotaxic Instruments, and allows replacing and centering the probe (for use only with ear bar at 0 coordinates), without removing the animal.
  • The hole is 30 mm above ear bar zero.