Learn More
Chemglass Life Sciences Needle, Syringe, Removable, 26s Gauge, .018in OD x .004in ID x 2.5in Long
SDP
Catalog No. CG307502
Encompass_Preferred
Needle, Syringe, Removable, 26s Gauge,.018in OD x .004in ID x 2.5in Long. Needle for CG-3070 and CG-3074 syringes. All needles are made of #304 full hard stainless steel and have a 22¿ beveled point for septum penetration along with general use. Removable needles come complete with the PTFE ferrule necessary for assembly.
Catalog No. CG307502 Supplier Chemglass Life Sciences Supplier No. CG307502
Needle, Syringe, Removable, 26s Gauge,.018in OD x .004in ID x 2.5in Long. Needle for CG-3070 and CG-3074 syringes. All needles are made of #304 full hard stainless steel and have a 22¿ beveled point for septum penetration along with general use. Removable needles come complete with the PTFE ferrule necessary for assembly.