Learn More
GA International Removable Autoclave-Resistant Labels for Laser Printers 2.63" x 1" / 66.8mm x 25.4mm Rectangle White From -40°C to +150°C (-40°F to +302°F)
Catalog No. NC1686053
Encompass_Preferred
Removable laser printable labels, ideal for identifying surgical instruments, metal and plastic sterilization containers, glassware, trays, blister packs, and other items intended for use in steam autoclaves, ovens, Ethylene oxide (EtO) sterilizers, as well as gamma irradiators. Water-proof and alcohol-resistant, they have been tested to endure one standard autoclave cycle (+121°C/+250°F, 16.5 psi, 20 minutes). Perfect for identifying one-use packages or reusable containers, as these durable autoclave-resistant laser labels will not leave any adhesive residue behind after removal. Compatible with all major brands of desktop laser printers, they are easy to format with our Microsoft Word free online templates.*Adhesive residue may remain after dry heat treatment exceeding 1 hour.
Catalog No. NC1686053 Supplier GA International Supplier No. ALTR3WH
Add to Cart
Add to Cart
Removable laser printable labels, ideal for identifying surgical instruments, metal and plastic sterilization containers, glassware, trays, blister packs, and other items intended for use in steam autoclaves, ovens, Ethylene oxide (EtO) sterilizers, as well as gamma irradiators. Water-proof and alcohol-resistant, they have been tested to endure one standard autoclave cycle (+121°C/+250°F, 16.5 psi, 20 minutes). Perfect for identifying one-use packages or reusable containers, as these durable autoclave-resistant laser labels will not leave any adhesive residue behind after removal. Compatible with all major brands of desktop laser printers, they are easy to format with our Microsoft Word free online templates.*Adhesive residue may remain after dry heat treatment exceeding 1 hour.