Learn More
New England Biolabs, Inc. Endo D – 1500 units
SDP
Catalog No. 50591178
Encompass
Endo D is a recombinant glycosidase, which cleaves within the chitobiose core of paucimannose N-linked glycans, with or without extensions in the antennae. Endo D is tagged with a chitin binding domain (CBD) for easy removal from a reaction, and is supplied glycerol-free for optimal performance in HPLC and MS- intensive methods.
Catalog No. 50-591-178 Supplier New England Biolabs, Inc. Supplier No. P0742S
Endo D is a recombinant glycosidase, which cleaves within the chitobiose core of paucimannose N-linked glycans, with or without extensions in the antennae. Endo D is tagged with a chitin binding domain (CBD) for easy removal from a reaction, and is supplied glycerol-free for optimal performance in HPLC and MS- intensive methods.