Learn More
New England Biolabs, Inc. Endo S – 30000 units
SDP
Catalog No. NC0520225
Encompass
Endo S is an endoglycosidase specific for cleaving the N-linked glycans from the chitobiose core of the heavy chain of native IgG. Endo S is tagged with a chitin binding domain (CBD) for easy removal from a reaction and is supplied glycerol free for optimal performance in HPLC and MS intensive methods.
Catalog No. NC0520225 Supplier New England Biolabs, Inc. Supplier No. P0741L
Endo S is an endoglycosidase specific for cleaving the N-linked glycans from the chitobiose core of the heavy chain of native IgG. Endo S is tagged with a chitin binding domain (CBD) for easy removal from a reaction and is supplied glycerol free for optimal performance in HPLC and MS intensive methods.