Learn More
New England Biolabs, Inc. Remove-iT PNGase F – 33750 units
SDP
Catalog No. NC0471659
Encompass
Remove-iT PNGase F is a tagged version of PNGase F, optimized for easy removal from a reaction. Remove-iT PNGase F is an amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides from N-linked glycoproteins.
Catalog No. NC0471659 Supplier New England Biolabs, Inc. Supplier No. P0706L
Remove-iT PNGase F is a tagged version of PNGase F, optimized for easy removal from a reaction. Remove-iT PNGase F is an amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides from N-linked glycoproteins.