Learn More
MSC AbilityOne Toxic Gases and Vapors Remover, Unscented, 5 gal, Powder
Catalog No. 502837037
Encompass
BoostR Toxic Gases and Vapors (VOCs) Remover scavenges reactive gases and vapors like hydrogen sulfide, sulfur dioxide, carbon monoxide, and solvents. It's for confined spaces, odor removal, corrosion control, and HAZMAT response. The product works in sewers, in air and water phases, and is simple, safe, and requires no prep or additives.
  • Scavenges reactive gases and vapors (hydrogen sulfide, sulfur dioxide, carbon monoxide, free hydrogen and oxygen, solvents).
  • Removes other sewer gases and noxious odors.
  • Suitable for confined safe spaces.
  • Aids in corrosion control and HAZMAT response/recovery.
  • Works in sewers in both air and water phases.
  • Simple and easy to use, skin and eye safe.
  • No prep, additives, or shelf-life needed.
  • Unscented, acid-free, phosphate-free, and PVC-free.
Catalog No. 50-283-7037 Supplier MSC Supplier No. 84138817
BoostR Toxic Gases and Vapors (VOCs) Remover scavenges reactive gases and vapors like hydrogen sulfide, sulfur dioxide, carbon monoxide, and solvents. It's for confined spaces, odor removal, corrosion control, and HAZMAT response. The product works in sewers, in air and water phases, and is simple, safe, and requires no prep or additives.
  • Scavenges reactive gases and vapors (hydrogen sulfide, sulfur dioxide, carbon monoxide, free hydrogen and oxygen, solvents).
  • Removes other sewer gases and noxious odors.
  • Suitable for confined safe spaces.
  • Aids in corrosion control and HAZMAT response/recovery.
  • Works in sewers in both air and water phases.
  • Simple and easy to use, skin and eye safe.
  • No prep, additives, or shelf-life needed.
  • Unscented, acid-free, phosphate-free, and PVC-free.