Learn More
Med Vet International Lotion Remover 3M Liquid .5 oz, 8610, 20/Box
Catalog No. 502830771
Encompass_Preferred
3M™ Remover lotion makes clean-up after surgery easier by dissolving DuraPrep™ film, tincture of benzoin and iodophor color It works quickly and easily, wiping away clean in under 30 seconds with no scrubbing needed Fragrance free so it's safe for sensitive skin Moisturizers leave skin feeling soft as well as clean Remover lotion will not impact the adhesion of tape or dressing, so bandages stay in place
Catalog No. 50-283-0771 Supplier Med Vet International Supplier No. 229914BX
3M™ Remover lotion makes clean-up after surgery easier by dissolving DuraPrep™ film, tincture of benzoin and iodophor color It works quickly and easily, wiping away clean in under 30 seconds with no scrubbing needed Fragrance free so it's safe for sensitive skin Moisturizers leave skin feeling soft as well as clean Remover lotion will not impact the adhesion of tape or dressing, so bandages stay in place