Learn More
Med Vet International Lotion Remover 3M Liquid .5 oz, 8610, 100/Case
Catalog No. 502830772
Encompass_Preferred
3M™ Remover lotion makes clean-up after surgery easier by dissolving DuraPrep™ film, tincture of benzoin and iodophor color It works quickly and easily, wiping away clean in under 30 seconds with no scrubbing needed Fragrance free so it's safe for sensitive skin Moisturizers leave skin feeling soft as well as clean Remover lotion will not impact the adhesion of tape or dressing, so bandages stay in place
Catalog No. 50-283-0772 Supplier Med Vet International Supplier No. 229914CS
3M™ Remover lotion makes clean-up after surgery easier by dissolving DuraPrep™ film, tincture of benzoin and iodophor color It works quickly and easily, wiping away clean in under 30 seconds with no scrubbing needed Fragrance free so it's safe for sensitive skin Moisturizers leave skin feeling soft as well as clean Remover lotion will not impact the adhesion of tape or dressing, so bandages stay in place