Learn More
Med Vet International Remover Lotion 3M Liquid 4 oz, 8611, 12/Case
Catalog No. 502831862
Encompass_Preferred
Removes iodophor color, tincture of benzoin, prep film and other materials soluble in alcohol No scrubbing necessary Wipes away quickly and easily in as little as 25 seconds Does not compromise tape or dressing adhesion Contains non-irritating and fragrance-free skin softeners and a moisturizer
Catalog No. 50-283-1862 Supplier Med Vet International Supplier No. 223357CS
Removes iodophor color, tincture of benzoin, prep film and other materials soluble in alcohol No scrubbing necessary Wipes away quickly and easily in as little as 25 seconds Does not compromise tape or dressing adhesion Contains non-irritating and fragrance-free skin softeners and a moisturizer