Learn More
TARGETMOL CHEMICALS INC REMOXIPRIDE HYDROCHLORID 5MG
SDP
Catalog No. 502580886
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Remoxipride hydrochloride (Roxiam) is a selective antagonist of D2 receptor with IC50s of 1.57 uM >100 uM and 42 uM for D2 receptor D1 receptor and (alpha)1-adrenoceptor. purity: 99%
Catalog No. 50-258-0886 Supplier TARGETMOL CHEMICALS INC Supplier No. T23232
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Remoxipride hydrochloride (Roxiam) is a selective antagonist of D2 receptor with IC50s of 1.57 uM >100 uM and 42 uM for D2 receptor D1 receptor and (alpha)1-adrenoceptor. purity: 99%