Learn More
MSC Tape Logic Laser Labels, Removable, Yellow, 3000/Case
Catalog No. 501913342
Encompass
These removable laser labels are designed for creating temporary labels for various projects. They measure 2 5/8 in. x 1 in. and are fluorescent yellow. The labels can be cleanly removed without leaving residue.
  • Lifts cleanly without residue
  • Blank for custom printing
  • Acid-free paper material
  • Rectangle shape with round corners
  • Fluorescent yellow color
  • Adhesive: Acrylic
  • Shelf life: 1 to 2 years (stored at room temperature)
  • Temperature performance: -4°F to 248°F
  • Meets FDA 21 CFR 175.105 for indirect food contact
  • Case contains 3000 labels.
Catalog No. 50-191-3342 Supplier MSC Supplier No. 39527833
These removable laser labels are designed for creating temporary labels for various projects. They measure 2 5/8 in. x 1 in. and are fluorescent yellow. The labels can be cleanly removed without leaving residue.
  • Lifts cleanly without residue
  • Blank for custom printing
  • Acid-free paper material
  • Rectangle shape with round corners
  • Fluorescent yellow color
  • Adhesive: Acrylic
  • Shelf life: 1 to 2 years (stored at room temperature)
  • Temperature performance: -4°F to 248°F
  • Meets FDA 21 CFR 175.105 for indirect food contact
  • Case contains 3000 labels.