Learn More
Electron Microscopy Sciences CultureWell™ Removeable Chambered Coverglass 16 well
SDP
Catalog No. 5034393
Encompass_Preferred
CultureWell with removable chambered coverglass is ideal for cell culture and fluorescence imaging applications. The 2 x 8, 16 chambered coverglass is like a chambered slide with the imaging advantages of a removable coverglass substructure. It is designed with standard 96 well plate volumes and well spacing. The non-cytotoxic silicone well gasket forms a leak-proof seal between the polystyrene upper structure and the coverglass. CultureWell Chambered Coverglass is shipped sterile and ready to use.
Catalog No. 50-343-93 Supplier Electron Microscopy Sciences Supplier No. 7045908
CultureWell with removable chambered coverglass is ideal for cell culture and fluorescence imaging applications. The 2 x 8, 16 chambered coverglass is like a chambered slide with the imaging advantages of a removable coverglass substructure. It is designed with standard 96 well plate volumes and well spacing. The non-cytotoxic silicone well gasket forms a leak-proof seal between the polystyrene upper structure and the coverglass. CultureWell Chambered Coverglass is shipped sterile and ready to use.